Learn More
Invitrogen™ Mouse IL-10 (ELISA Standard) Recombinant Protein
Description
Description: mouse IL-10 is an ~18 kDa cytokine, also known as Cytokine Synthesis Inhibitory Factor (CSIF). In the mouse, Th2 cells, B1 cells and keratinocytes are the major cell subsets that produce IL-10. IL-10 inhibits synthesis of Th1 cytokines and proliferation of T cells, and acts as a costimulatory signal for mast cells, developing thymocytes and the Th2 response. Applications Reported: The Mouse IL-10 Lyophilized Standard is intended to be used as a standard for the Mouse IL-10 Ready-Set-Go! ELISA kit (Product # 88-7105). Typically the standard is run in a series of 7 doubling dilutions from the 'top' standard. Please refer to the Certificate of Analysis included with each ELISA set for instructions on the proper reconstitution and dilution of the lyophilized standard. Note: These are single-use vials. Use one time within 4 hours of reconstitution and discard. Reconstitute and dilute as suggested in the Certificate of Analysis or recovery may not be accurate. Applications Tested: The Mouse IL-10 Lyophilized Standard has been calibrated and tested in the Mouse IL-10 Ready-Set-Go! ELISA kit (Product # 88-7105-76). Source: E. coli expressed amino acids Ser 19-Ser 178 of mature mouse IL-10 accession # NM_010548 with a Cys149tyr substitution.
Specifications
Specifications
| Accession Number | P18893 |
| For Use With (Application) | Neutralization, ELISA Standard |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 16153 |
| Molecular Weight (g/mol) | 18 kDa |
| Name | Mouse IL-10 (ELISA Standard) |
| Purification Method | Purified |
| Quantity | 4 pack |
| Source | E. Coli |
| Storage Requirements | 4°C |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.