Learn More
ACROBiosystems Mouse IL-10 R alpha / CD210 Protein, Fc Tag
Catalog No. 502331080
Encompass_Preferred
Mouse IL-10 R alpha Protein, Fc Tag (ILR-M5254) is expressed from human 293 cells (HEK293). It contains AA Leu 17 - Ile 241 (Accession # Q61727).
Catalog No. 50-233-1080 Supplier ACROBiosystems Supplier No. ILRM5254100UG
Mouse IL-10 R alpha Protein, Fc Tag (ILR-M5254) is expressed from human 293 cells (HEK293). It contains AA Leu 17 - Ile 241 (Accession # Q61727).