Learn More
Kingfisher Biotech Inc Mouse IL-13 Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0735M025
Encompass_Preferred
The Mouse IL-13 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Mouse IL-13 applications are for cell culture, ELISA standard, and Western Blot Control. The Mouse IL-13 yeast-derived recombinant protein can be purchased in multiple sizes. Mouse IL-13 Specifications: (Molecular Weight: 12.2 kDa) (Amino Acid Sequence: GPVPRSVSLP LTLKELIEEL SNITQDQTPL CNGSMVWSVD LAAGGFCVAL DSLTNISNCN AIYRTQRILH GLCNRKAPTT VSSLPDTKIE VAHFITKLLS YTKQLFRHGP F (111)) (Gene ID: 16163). For research use only. Made in the USA
Catalog No. 50-241-3465