Learn More
Raybiotech Inc Mouse IL-13 ELISA Kit
Supplier:  Raybiotech Inc ELMIL131
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 6 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA kit shows no cross-reactivity with any of the cytokines tested: Mouse 6Ckine, CTACK, Eotaxin, GCSF, GM-CSF, IL-2, IL-3, IL-4, IL-5, IL-6, IL-9, IL-10, IL-12p40, IL-12p70, IL-17, IFN-gamma, KC, Leptin, MCP-5, MIP-1 alpha, MIP-2, MIP-3 beta, RANTES, SCF, sTNFri, TARC, TIMP-1, TNF-alpha, TPO, VEGF.
Catalog No. 50-195-0033