Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse IL-15 Recombinant Protein, PeproTech®

Catalog No. 210152UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 μg
2 x 500 μg
250 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
210-15-2UG 2 μg
21-015-01M 2 x 500 μg
21015100UG 100 μg
210-15-10UG 10 μg
21015250UG 250 μg
21015500UG 500 μg
210-15-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 210-15-2UG Supplier Gibco™ Supplier No. 210152UG
  • $92.25
    $78.65 / Each of 1

    Save $13.60

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recombinant Protein

210-15-1MG will be provided as 2 x 500 μg (210-15-500UG). Recombinant Murine IL-15 is a 13.3 kDa protein consisting of 115 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis.

IL-15 (Interleukin 15 ) is a cytokine that regulates T and natural killer cell activation and proliferation. IL-15 and IL-2 share many biological activities as both have been found to bind common hematopoietin receptor subunits, and may compete for the same receptor, and thus negatively regulate each other's activity. The number of CD8+ memory cells is shown to be controlled by a balance between IL-15 and IL-2. IL-15 induces the activation of JAK kinases, as well as the phosphorylation and activation of transcription activators STAT3, STAT5, and STAT6. In mouse, studies suggest that IL-15 may increase the expression of apoptosis inhibitor Bcl-xL, possibly through the transcription activation activity of STAT6, and thus prevent apoptosis. IL-15 plays an important role in the growth and differentiation of T and B lymphocytes, natural killer cells, macrophages, and monocytes as well as activation of a number of important intracellular signaling molecules. As such, IL-15 could be essential for the immune responses, allograft rejection, and the pathogenesis of autoimmune diseases. Further, IL-15 is a widely expressed pro-inflammatory cytokine and has been shown to play a role in several inflammatory disorders, including rheumatoid arthritis, psoriasis and pulmonary inflammatory diseases. Emerging data suggest that IL-15 may serve as a good therapeutic target, as there appears to be a beneficial effect of IL-15 neutralization in models of psoriasis and diabetes.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P48346
For Use With (Application) Control, Western Blot Control, ELISA Standard, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 16168
Molecular Weight (g/mol) 13.3 kDa
Name Mouse IL-15
Purification Method Purified
Quantity 2 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias AI503618; il 15; Il15; IL-15; ILN; Interleukin; interleukin 15; Interleukin15; interleukin-15; MGC9721; M-IL-15
Common Name IL-15
Gene Symbol IL15
Biological Activity The ED50 was determined by the stimulation of the proliferation of murine CTLL-2 cells is ≤ 5.0 ng/ml, corresponding to a specific activity of ≥ 2 x 10^5 units/mg
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence MNWIDVRYDL EKIESLIQSI HIDTTLYTDS DFHPSCKVTA MNCFLLELQV ILHEYSNMTL NETVRNVLYL ANSTLSSNKN VAESGCKECE ELEEKTFTEF LQSFIRIVQM FINTS
Content And Storage -20°C
Activity The ED50 was determined by the stimulation of the proliferation of murine CTLL-2 cells is ≤ 5.0 ng/ml, corresponding to a specific activity of ≥ 2 x 10^5 units/mg.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.