Learn More
FUJIFILM BIOSCIENCES INC Mouse IL-16, Recombinant Protein, Shenandoah Biotechnology, 100 ug vial
Supplier:  FUJIFILM BIOSCIENCES INC 20049100UG
Encompass_Preferred
Interleukin 16 (IL-16) is produced by CD4+ and CD8+ T cells and functions as a chemoattractant for lymphocytes, monocytes, eosinophils, dendritic cells, and Langerhans cells.  Additionally, IL-16 upregulates the expression of the interleukin 2 receptor (CD25), modulates T lymphocyte growth, and suppresses the replication of the human immunodeficiency virus (HIV) and simian immunodeficiency viruses (SIV).
Catalog No. 50-990-886