Learn More
Kingfisher Biotech Inc Mouse IL-17A Recombinant Protein (Yeast, Endotoxin - Free) - 100 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0355M100
Encompass_Preferred
The Mouse IL-17A yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Mouse IL-17A applications are for cell culture, ELISA standard, and Western Blot Control. The Mouse IL-17A yeast-derived recombinant protein can be purchased in multiple sizes. Mouse IL-17A Specifications: (Molecular Weight: 15.0 kDa) (Amino Acid Sequence: AAIIPQSSAC PNTEAKDFLQ NVKVNLKVFN SLGAKVSSRR PSDYLNRSTS PWTLHRNEDP DRYPSVIWEA QCRHQRCVNA EGKLDHHMNS VLIQQEILVL KREPESCPFT FRVEKMLVGV GCTCVASIVR QAA (133)) (Gene ID: 16171). For research use only. Made in the USA
Catalog No. 50-241-3515