Learn More
Raybiotech Inc Mouse IL-17 RB ELISA Kit
Supplier:  Raybiotech Inc ELMIL17BR1
Encompass_Preferred
For serum, plasma, and cell culture supernatant samples. Sensitivity: 0.2 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: Mouse Adiponectin (ACRP30), ANGPTL1, ANGPTL3, Artemin, CCL28 (MEC), CD36 (SR-B3), Chordin, CRP, E-Cadherin, Epigen, Epiregulin, Fas, Galectin-7, gp130, Granzyme B, Gremlin-1, IFN-gamma R1, IL-17B, IL-22, MIP-1beta, MMP-10, MMP-2, MMP-3, PDGF-AA, Persephin, sFRP-3, SLAM, Shh-N, TCK-1, TECK, TGF beta 1, Trance, TREML1, TWEAK, VEGF-B, VEGFR2
Catalog No. 50-195-0037