Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse IL-18 BPc ELISA Kit

Catalog No. EM43RBX10
Encompass_Preferred
Change view
Click to view available options
10 x 96 Tests; HRP
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
EM43RBX10 10 x 96 Tests; HRP
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EM43RBX10 Supplier Invitrogen™ Supplier No. EM43RBX10
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

ELISA

Mouse IL-18 BPc quantitates mouse IL-18 BPc in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse IL-18 BPc.

The IL18BPc gene encodes one of the isoforms of the interleukin-18 binding protein (IL-18BP), a crucial component in immune system regulation. Located in humans, IL18BPc is among four isoforms, alongside hIL-18BPa, b, and d, with hIL-18BPa being the predominant form. Both hIL-18BPa and hIL-18BPc effectively neutralize interleukin-18 (IL-18), a pro-inflammatory cytokine produced by T cells and NK cells, thereby mitigating its activity. This interaction is essential as IL-18 is implicated in several inflammatory and cardiovascular diseases. IL-18BP acts as a natural inhibitor, demonstrating a high affinity for IL-18, which makes it a significant focus for therapies targeting cytokine pathways in immunotherapy. Its ability to antagonize IL-18 highlights its potential as an immune checkpoint in cancer therapy, offering alternatives to enhance treatment efficacy and tolerability.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 22-5000 pg/mL
Assay Sensitivity 22 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Mouse
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias IL18 BPC; IL18BP; IL-18BP; IL-18BPC; Interleukin 18 binding proteinIL18 BPC; IL18BP; IL-18BP; IL-18BPC; Interleukin 18 binding protein;
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target IL18BPc
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.