Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse IL-18 Uncoated ELISA Kit with Plates

Catalog No. 885061886
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
88-506-1886 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 88-506-1886 Supplier Invitrogen™ Supplier No. 885061886
Add to Cart
Add to Cart

ELISA

The protein encoded by this gene is a proinflammatory cytokine. This cytokine can induce the IFN-gamma production of T cells. The combination of this cytokine and IL12 has been shown to inhibit IL4 dependent IgE and IgG1 production, and enhance IgG2a production of B cells. IL-18 binding protein can specifically interact with this cytokine, and thus negatively regulate its biological activity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P70380
Assay Range 31.2-2000 pg/mL
Assay Sensitivity <31.2 pg/mL
Conjugate HRP
Immunoassay Kit Format Uncoated ELISA Kit with Plates
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Mouse
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias cytokine; iboctadekin; IFN-gamma-inducing factor; IGIF; il 18; IL-1 gamma; IL18; IL-18; IL1F4; IL-1g; ILN; interferon gamma inducing factor; interferon gamma-inducing factor; interferon-gamma-inducing factor; Interleukin; interleukin 18; interleukin 18 (interferon-gamma-inducing factor); interleukin-1 gamma; Interleukin18; Interleukin-18; interleukin-18 proprotein; interleukin-18-like protein; MGC12320cytokine; iboctadekin; IFN-gamma-inducing factor; IGIF; il 18; IL-1 gamma; IL18; IL-18; IL1F4; IL-1g; ILN; interferon gamma inducing factor; interferon gamma-inducing factor; interferon-gamma-inducing factor; Interleukin; interleukin 18; interleukin 18 (interferon-gamma-inducing factor); interleukin-1 gamma; Interleukin18; Interleukin-18; interleukin-18 proprotein; interleukin-18-like protein; MGC12320;
Gene ID (Entrez) 16173
Gene Symbol IL18
Kit Contents Capture Antibody: Pre-titrated, purified antibody, Detection Antibody: Pre-titrated, HRP-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22, -76, -86), Purified antibody, Detection Antibody: Pre-titrated, HRP-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22, -76, -86)
Label or Dye Biotin
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet Ice
Storage Requirements 2-8°C
Target IL-18
Test Time 1 hr 45 min
Total Assay Time 24 hr 30 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.