Learn More
Sino Biological Mouse IL-2 ELISpot Kit (ALP) 10 Kit (96 Tests)
Catalog No. 5000696253
Encompass_Preferred
The kit applies for the detection of mouse IL-2 at the cellular level by counting IL-2-secreting cells.
Catalog No. 50-006-96253 Supplier Sino Biological Supplier No. EK51061A10
The kit applies for the detection of mouse IL-2 at the cellular level by counting IL-2-secreting cells.