Learn More
FUJIFILM BIOSCIENCES INC Mouse IL-2, Recombinant Protein, Shenandoah Biotechnology, 5 ug vial
Supplier:  FUJIFILM BIOSCIENCES INC 200175UG
Encompass_Preferred
Interleukin 2 (IL-2) is an immunomodulatory cytokine that is produced by lymphocytes.  IL-2 signals through the IL-2R receptor to induce activated T cell proliferation and promote T cell differentiation. IL-2 also stimulates the proliferation and differentiation of B cells, natural killer cells, monocytes, and macrophages.
Catalog No. 50-990-845
Edge