Learn More
ACROBiosystems ACROBiosystems Mouse IL-2 Protein, His Tag
Supplier:  ACROBiosystems IL2M52H3100UG
Encompass_Preferred
Recombinant Protein;100UG;Mouse IL-2, His Tag (IL2-M52H3) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Gln 169 (Accession # P04351-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 19.1 kDa. The protein migrates as 20-25 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse IL-2, His Tag (Cat. No. IL2-M52H3) at 5 μg/mL (100 μL/well) can bind Mouse IL-2RB&IL-2RA&IL-2RG, Fc Tag&Fc Tag (Cat. No. ILG-M5253) with a linear range of 0.8-3 ng/mL (QC tested).
Catalog No. 50-210-6500