Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mouse IL-2 Quantikine ELISA Kit
SDP

Catalog No. SM2000
Change view
Click to view available options
Quantity:
1 Kit
1 Kit (1 Plate)
1 Pack
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SM2000 1 Pack
M20001 1 Kit (1 Plate)
M2000 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. SM2000 Supplier R&D Systems™ Supplier No. SM2000

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse IL-2 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse IL-2 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse IL-2 in cell culture supernates (50 ul), serum (50 ul).

Specifications

Assay Range 15.60 to 1000 pg/mL
Assay Sensitivity 3 pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Immunoassay Kit Format 96-well Plate
Product Line R&D Systems
Sample Type Cell Culture Supernatant, Serum
Specificity Natural and Recombinant Mouse IL-2
Target Species Mouse
For Use With (Application) ELISA
Gene ID (Entrez) 16183
Quantity 1 Pack
Sample Volume Cell Culture Supernates (50uL),Serum (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym aldesleukin, IL2, IL-2 lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, T cell growth factor, T-cell growth factor, TCGF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.