Learn More
ACROBiosystems ACROBiosystems Mouse IL-2 R beta&IL-2 R alpha&IL-2 R gamma Protein, Fc Tag&Fc Tag
Supplier:  ACROBiosystems ILGM5253100UG
Encompass_Preferred
Recombinant Protein;100UG;Mouse IL-2RB&IL-2RA&IL-2RG, Fc Tag&Fc Tag (ILG-M5253) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Glu 240 & Glu 22 - Lys 236 & Trp 23 - Ala 263 (Accession # P16297-1 (IL-2RB) & P01590-1 (IL-2RA) & P34902-1 (IL-2RG)).;Mouse IL-2RB&IL-2RA&IL-2RG, Fc Tag&Fc Tag is produced by co-expression of IL-2RB and IL-2RA and IL-2RG, has a calculated MW of 51.5 kDa (IL-2RB) and 79.4 kDa (IL-2RA&IL-2RG). Subunit IL-2RB is fused with a human IgG1 Fc tag at the C-terminus and subunit IL-2RA&IL-2RG is fused with a human IgG1 Fc tag at the C-terminus. The reducing (R) protein migrates as 75-90 kDa and 120-150 kDa respectively due to glycosylation.;Immobilized Human IL-2, Tag Free (Cat. No. IL2-H4113) at 5 μg/mL (100 μL/well) can bind Mouse IL-2RB&IL-2RA&IL-2RG, Fc Tag&Fc Tag (Cat. No. ILG-M5253) with a linear range of 0.05-0.313 μg/mL (QC tested).
Catalog No. 50-210-7258