Learn More
ACROBiosystems ACROBiosystems Mouse IL-2 R beta&IL-2 R gamma Heterodimer Protein, His Tag&Twin-Strep Tag (MALS verified)
Supplier:  ACROBiosystems ILGM52831MG
Encompass_Preferred
Recombinant Protein;1MG;Mouse IL-2RB&IL-2RG Heterodimer Protein, His Tag&Twin-Strep Tag (ILG-M5283) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Glu 240 (IL-2RB) & Trp 23 - Ala 263 (IL-2RG) (Accession # P16297-1 (IL-2RB) & P34902-1 (IL-2RG)).;Mouse IL-2RB&IL-2RG Heterodimer Protein, His Tag&Twin-Strep Tag is produced by co-expression of IL-2RB and IL-2RG, has a calculated MW of 30.4 kDa (IL-2RB) and 35.0 kDa (IL-2RG). Subunit IL-2RB is fused with a polyhistidine tag at the C-terminus and subunit IL-2RG is fused with Twin-Strep tag at the C-terminus. The reducing (R) protein migrates as 50-60 kDa and 62-66 kDa respectively due to glycosylation.
Catalog No. 50-210-7117