Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse IL-20 R beta ELISA Kit

Catalog No. EM45RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EM45RBX5 5 x 96 Tests
EM45RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EM45RBX5 Supplier Invitrogen™ Supplier No. EM45RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

ELISA

Mouse IL-20 R beta quantitates mouse IL-20 R beta in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse IL-20 R beta.

IL20RB and IL20RA (MIM 605620) form a heterodimeric receptor for interleukin-20 (IL20; MIM 605619) (Blumberg et al., 2001 [PubMed 11163236]).[supplied by OMIM, Feb 2009].
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AV228068; DIRS1; Fibronectin type III domain containing 6; FNDC6; Gm186; IL-20 receptor subunit beta; IL20R2; IL-20R2; IL20RB; IL-20 RB; IL-20 R-beta; interleukin 20 receptor beta; interleukin 20 receptor beta subunit; interleukin 20 receptor subunit beta; interleukin-20 receptor II; interleukin-20 receptor subunit beta; RGD1566151; UNQ557/PRO1114
Assay Range 0.45-100 ng/mL
Assay Sensitivity 0.45 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Mouse
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias AV228068; DIRS1; Fibronectin type III domain containing 6; FNDC6; Gm186; IL-20 receptor subunit beta; IL20R2; IL-20R2; IL20RB; IL-20RB; IL-20R-beta; interleukin 20 receptor beta; interleukin 20 receptor beta subunit; interleukin 20 receptor subunit beta; interleukin-20 receptor II; interleukin-20 receptor subunit beta; RGD1566151; UNQ557/PRO1114AV228068; DIRS1; Fibronectin type III domain containing 6; FNDC6; Gm186; IL-20 receptor subunit beta; IL20R2; IL-20R2; IL20RB; IL-20RB; IL-20R-beta; interleukin 20 receptor beta; interleukin 20 receptor beta subunit; interleukin 20 receptor subunit beta; interleukin-20 receptor II; interleukin-20 receptor subunit beta; RGD1566151; UNQ557/PRO1114;
Gene ID (Entrez) 213208
Gene Symbol Il20rb
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target IL20RB
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.