Learn More
Raybiotech Inc Mouse IL-20 R beta ELISA Kit
Supplier:  Raybiotech Inc ELMIL20RB1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.45 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Mouse IL-20 R beta.
Catalog No. 50-195-0046