Learn More
Kingfisher Biotech Inc Mouse IL-22 Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0903M025
Encompass_Preferred
The Mouse IL-22 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Mouse IL-22 applications are for cell culture, ELISA standard, and Western Blot Control. The Mouse IL-22 yeast-derived recombinant protein can be purchased in multiple sizes. Mouse IL-22 Specifications: (Molecular Weight: 16.7 kDa) (Amino Acid Sequence: LPVNTRCKLE VSNFQQPYIV NRTFMLAKEA SLADNNTDVR LIGEKLFRGV NAKDQCYLMK QVLNFTLEDV LLPQSDRFQP YMQEVVPFLT KLSNQLSSCH ISGDDQNIQK NVRRLKETVK KLGESGEIKA IGELDLLFMS LRNACV (146)) (Gene ID: 50929). For research use only. Made in the USA
Catalog No. 50-241-3625