Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse IL-23 (Carrier-Free) Recombinant Protein, eBioscience™

Catalog No. 509179
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-917-9 500 μg
50-112-3500 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-917-9 Supplier Invitrogen™ Supplier No. 34823185
Only null left
Add to Cart
Add to Cart
Request a Quote

Recombinant Protein

IL-23 is a heterodimeric cytokine composed of the p40 subunit of IL-12 disulfide-linked with a protein p19. p19, like p35 of IL-12, is biologically inactive by itself. IL-23 interacts with IL-12Rbeta1 and an additional, novel beta2-like receptor subunit with STAT4 binding domain, termed IL-23R. IL-23 is secreted by activated mouse and human dendritic cells. Biological activities of mouse IL-23 are distinct from those of mouse IL-12. Mouse IL-23 was found not to induce significant amounts of IFN-γ. Mouse IL-23 does induce strong proliferation of memory T cells (but not naive T cells), whereas IL-12 has no effect on memory cells. Additionally, mouse IL-23 (but not IL-12) can activate mouse memory T cells to produce the proinflammatory cytokine IL-17. Human IL-23 has biological properties which are less distinct from human IL-12; human IL-23 induces proliferation of memory T cells and induces moderate levels of IFN-γ production by naive and memory T cells, as compared to IL-12.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9EQ14
Concentration 0.5 mg/mL
For Use With (Application) Western Blot Control, Bioactivity, Western Blot
Formulation PBS with no preservative; pH 6.0
Gene ID (Entrez) 83430
Molecular Weight (g/mol) 55.6 kDa
Name Mouse IL-23 (Carrier-Free)
pH Range 6.0
Purification Method Purified
Quantity 500 μg
Source Insect cells
Storage Requirements -80°C
Regulatory Status RUO
Endotoxin Concentration <0.01 ng/μg
Gene Alias il 23; IL 23 A; IL12B; Il-12b; Il12p40; Il-12p40; il23; IL-23; IL-23 subunit alpha; IL23A; IL-23A; IL-23-A; IL23P19; IL-23p19; ILN; Interleukin; interleukin 12B; interleukin 23 p19 subunit; interleukin 23 subunit alpha; interleukin 23, alpha subunit p19; interleukin-23 subunit alpha; Interleukin-23 subunit p19; interleukin-six, G-CSF related factor; JKA3 induced upon T-cell activation; MGC79388; P19; p40; RP23-388G23.1; SGRF; UNQ2498/PRO5798
Common Name IL-23 p19
Gene Symbol IL23A
Biological Activity ED50 ≤ 625 pg/mL; determined by the dose-dependent induction of IL-17 A expression in mouse splenocytes. This corresponds to a specific actvity greater than or equal to 1.5 x 10e6 Units/mg.
Product Type Protein
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Sequence Mouse IL-23, p40 subunit amino acids Met1-Ser355 (Accession # NM_008352) was coexpressed with subunit p19 amino acids Met1-Ala196 (Accession # AF301619)
Content And Storage -80°C
Activity ED50 ≤ 625 pg/mL; determined by the dose-dependent induction of IL-17A expression in mouse splenocytes. This corresponds to a specific actvity greater than or equal to 1.5x10e6 Units/mg.
Expression System Insect cells
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade >98% by SDS-PAGE
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.