Learn More
Raybiotech Inc Mouse IL-23 p19 ELISA Kit
Supplier:  Raybiotech Inc ELMIL231
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 25 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Mouse interleukin 23 alpha subunit p19, IL-23p19, IL-23A, MGC79388, SGRF, IL23A.
Catalog No. 50-195-0050