Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse IL-27 Uncoated ELISA Kit with Plates

Catalog No. 5017305
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-05 2 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-05 Supplier Invitrogen™ Supplier No. 88727422
Add to Cart
Add to Cart

ELISA

The Mouse Interleukin 27 (IL-27) Uncoated ELISA Kit contains pre-matched antibody pairs, plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of mouse IL-27. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

IL-27, a member of the IL-12 family, is a heterodimeric protein consisting of the p40-related protein Epstein-Barr virus-induced gene 3 (EBI3) non-covalently linked to an IL-12p35-related protein, p28 (also known as IL-30). IL-27 is produced by activated APCs and mature dendritic cells. IL-27 exerts its activities on NK cells and naive CD4+ T cells; mRNA expression analysis of IL-27 receptor components (WSX-1/TCCR and gp130) suggests that IL-27 may also target other cells, including mast cells and monocytes. Binding of IL-27 to WSX-1/gp130 activates JAK1, STAT1, and STAT3 and STAT1/3 phosphorylation. WSX-1/TCCR-deficient mice develop impaired Th1 responses and are more susceptible to infection with L. monocytogenes suggesting that Th1 responses require IL-27. Although activation of WSX-1 is required for the initiation of Th1 responses, it is not necessary for maintaining Th1 responses. IL-27 alone is not able to induce the differentiation of CD4+ T cells into IFN-γ-producing cells, suggesting a role for IL-27 as an initial activator of Th1 responses. An important effect of IL-27 in initiating Th1 responses is the induction of the Th1-specific transcription factor T-bet as well as the suppression of the Th2-specific transcription factor GATA-3. T-bet plays a critical role in Th1 differentiation by its ability to maintain IL-12Rβ2 expression following CD4+ T cell activation. Recent studies indicate that IL-27 has a potent antitumor activity. In vitro, IL-27 has been found to act directly on naive CD8 cells, generating CTL with enhanced granzyme B expression. In vivo, IL-27 has been reported to augment CTL activity, inhibit tumor growth, and induce complete regression of primary and metastatic neuroblastoma tumors.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8K3I6
Assay Range 16-2000 pg/mL
Assay Sensitivity 16 pg/mL
Content And Storage Capture Antibody: Pre-titrated, purified antibody
Detection Antibody: Pre-titrated, biotin-conjugated antibody
Standard: Recombinant cytokine for generating standard curve and calibrating samples
Coating Buffer: 10X PBS ELISA Coating Buffer
Assay Diluent: 5X concentrated
Detection enzyme: Pre-titrated Avidin-HRP
Substrate Solution: Tetramethylbenzidine (TMB) Substrate Solution
Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards
96 Well Plate: Corning Costar 9018 (included with product Cat. Nos. ending in suffixes -22, -44, -76, -86)
Conjugate HRP
Immunoassay Kit Format Uncoated ELISA Kit with Plates
Product Type ELISA
Sample Type Serum, Supernatant
Target Species Mouse
For Use With (Application) ELISA
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias IL27; IL-27; IL-27 p28 subunit; IL-27 subunit alpha; Il27a; IL-27A; IL-27-A; IL27-A; IL27p28; IL-27p28; IL30; IL-30; ILN; Interleukin; interleukin 27; interleukin 30; Interleukin-27; interleukin-27 subunit alpha; Interleukin-30; p28IL27; IL-27; IL-27 p28 subunit; IL-27 subunit alpha; Il27a; IL-27A; IL-27-A; IL27-A; IL27p28; IL-27p28; IL30; IL-30; ILN; Interleukin; interleukin 27; interleukin 30; Interleukin-27; interleukin-27 subunit alpha; Interleukin-30; p28; C87042; IL-17F;Ctla-8; Ctla8; IL-17; IL-17A; Il17
Gene ID (Entrez) 246779
Gene Symbol Il27
Kit Contents Capture Antibody: Pre-titrated, purified antibody, Detection Antibody: Pre-titrated, HRP-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22, -76, -86), Purified antibody, Detection Antibody: Pre-titrated, HRP-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22, -76, -86)
Label or Dye Biotin
Protein Aliases IL-27 subunit alpha, IL-27-A, IL27-A, p28
Quantity 2 x 96 Tests
Regulatory Status RUO
Sample Volume Serum 100 μL, Supernatant 100 μL
Storage Requirements 2-8°C
Synonym Interleukin-27
Target IL-27 p28
Test Time 1 hr 45 min
Total Assay Time 24 hr 30 min
Show More Show Less
What are the critical parameters for successful ELISPOT assay?

A: It is critically important to use a high-affinity binding PVDF membrane plate, rather than regular ELISA plate or nitrocellulose membrane plate. Additionally, highest affinity antibodies for capture work best in this assay. Empirical optimization of conditions such as cell density, stimulus/mitogen, and kinetics is necessary.

I recently bought one of your Ready-Set-Go! ELISA Sets, can I use the antibody pairs to run an ELISPOT?

We do not recommend the use of our ELISA kits for the ELISPOT application. We cannot guarantee the successful use of our ELISA kits in an ELISPOT application.

Do your Ebioscience antibody pairs work in ELISPOT as well?

Most, but not all, of our ELISA antibody pairs work for ELISPOT. We continue to evaluate and optimize our anti-human and anti-mouse cytokine antibody pairs for ELISPOT applications. Please see our protocols page (www.thermofisher.com/us/en/home/references/protocols/cell-and-tissue-analysis/protocols) for a detailed protocol and a reference table with the list of antibody pairs that can be used in ELISPOT.

What are the components of the Ready-Set-Go! ELISPOT Sets?

Capture Antibody: Pre-titrated, Functional Grade (low endotoxin) purified antibody
Detection Antibody: Pre-titrated, biotin-conjugated antibody
10X Sterile Azide-Free Coating Buffer
5X ELISA/ELISPOT Diluent
Detection enzyme: pre-titrated Avidin HRP
Certificate of Analysis: Lot-specific instructions for dilution of antibodies and enzyme

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.