Learn More
FUJIFILM BIOSCIENCES INC Mouse p28 subunit (IL-27/p28), Recombinant Protein, Shenandoah Biotechnology, 10 ug vial
Supplier:  FUJIFILM BIOSCIENCES INC 2005010UG
Encompass_Preferred
The p28 subunit of interleukin 27 (IL-27), also known as interleukin 30 (IL-30), is a member of the interleukin 12 (IL-12) family of cytokines.  p28 is a secreted polypeptide that associates with the Epstein-Barr virus induced gene 3 (EBI3) to form the IL-27 cytokine heterodimer complex.  IL-27 functions as a proinflammatory cytokine that induces immunomodulatory effects in naïve CD4+ T cells, mast cells, and monocytes.  p28 can also form a complex with cytokine-like factor 1 (CLF), that is secreted by dendritic cells, to regulate natural killer (NK) and T cell functions. 
Catalog No. 50-990-889