Learn More
FUJIFILM BIOSCIENCES INC Mouse IL-3, Recombinant Protein, Shenandoah Biotechnology, 10 ug vial
Supplier:  FUJIFILM BIOSCIENCES INC 2000110UG
Encompass_Preferred
Interleukin 3 (IL-3) is a cytokine that is produced by activated T cells and mast cells.  IL-3 induces the differentiation of hematopoietic stem cells into myeloid precursor cells, such as erythrocyte, megakaryocyte, granulocyte, monocyte, and dendritic cells.  IL-3 also functions in the nervous system and is important during the B-1 cell regulation of chronic inflammatory diseases.  
Catalog No. 50-589-923