Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse IL-31 Recombinant Protein, PeproTech®

Catalog No. 21031100UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21031100UG 100 μg
210-31-10UG 10 μg
210-31-1MG 1 mg
21031250UG 250 μg
210-31-2UG 2 μg
21031500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 21031100UG Supplier Gibco™ Supplier No. 21031100UG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine IL-31 is a 15.8 kDa protein containing 141 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Interleukin-31 (IL-31) is a recently discovered T-cell cytokine closely related to IL-6 type cytokines and is preferentially produced by T helper type 2 cells. IL-31 activity is mediated through the ligand-induced oligomerization of a dimeric receptor complex containing IL-31 receptor A and oncostatin M receptor. In response to IL-31 binding, these proteins activate the JAK/STAT and the AKT signaling pathways. RNA levels of IL-31 receptor A and oncostatin M receptor are induced in activated monocytes but are expressed constitutively in epithelial cells. IL-31, when overexpressed in transgenic mice, results in the development of pruritis, alopecia, and skin lesions and in humans may result in atopic dermatitis, suggesting that IL-31 may represent a novel target for antipruritic drug development.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6EAL8
For Use With (Application) Control
Formulation Protein with no preservative
Gene ID (Entrez) 76399
Molecular Weight (g/mol) 15.8 kDa
Name Mouse IL-31
Purification Method Purified
Quantity 100 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias 1700013B14Rik; IL31; IL-31; ILN; Interleukin; interleukin 31; interleukin-31
Common Name IL-31
Gene Symbol Il31
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence MTCSLSFGAP ISKEDLRTTI DLLKQESQDL YNNYSIKQAS GMSADESIQL PCFSLDREAL TNISVIIAHL EKVKVLSENT VDTSWVIRWL TNISCFNPLN LNISVPGNTD ESYDCKVFVL TVLKQFSNCM AELQAKDNTT C
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.