Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse IL-36 beta/IL-1F8 ELISA Kit

Catalog No. EM46RBX10
Encompass_Preferred
Change view
Click to view available options
10 x 96 Tests; HRP
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
EM46RBX10 10 x 96 Tests; HRP
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EM46RBX10 Supplier Invitrogen™ Supplier No. EM46RBX10
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

ELISA

Mouse IL-36 beta/IL-1F8 quantitates mouse IL-36 beta/IL-1F8 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse IL-36 beta/IL-1F8.

IL-36B is is a member of the interleukin 1 cytokine family whose gene and eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. IL-36B is thought to activate the NF-kappaB pathway through IL-1 receptor family members IL-1RL2 and IL-1RAcP. Like the related proteins IL-36A and IL-36G, IL-36B requires post-translational processing for full agonist activity, but the cleavage mechanism is currently unknown. The IL-36 cytokines have been suggested to amplify Th1 responses by enhancing proliferation and Th1 polarization of naive CD4+ T cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9D6Z6
Assay Range 6.5-1500 pg/mL
Assay Sensitivity 6.5 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Mouse
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 2310043N20Rik; family of interleukin 1-eta; FIL1; FIL1 eta; FIL1-(ETA); Fil1e; FIL1H; FILI-(ETA); IL-1 eta; IL1-ETA; IL1F8; IL-1F8; IL1F8 (Canonical product IL-1F8a); IL-1F8 (FIL1-eta); IL1H2; IL-1H2; Il36b; Interleukin; interleukin 1 family, member 8; interleukin 1 family, member 8 (eta); interleukin 1, eta; interleukin 36, beta; interleukin-1 eta; Interleukin-1 family member 8; interleukin-1 homolog 2; Interleukin-1 Superfamily e; interleukin-36 beta2310043N20Rik; family of interleukin 1-eta; FIL1; FIL1 eta; FIL1-(ETA); Fil1e; FIL1H; FILI-(ETA); IL-1 eta; IL1-ETA; IL1F8; IL-1F8; IL1F8 (Canonical product IL-1F8a); IL-1F8 (FIL1-eta); IL1H2; IL-1H2; Il36b; Interleukin; interleukin 1 family, member 8; interleukin 1 family, member 8 (eta); interleukin 1, eta; interleukin 36, beta; interleukin-1 eta; Interleukin-1 family member 8; interleukin-1 homolog 2; Interleukin-1 Superfamily e; interleukin-36 beta;
Gene ID (Entrez) 69677
Gene Symbol Il1f8
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target IL36B
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.