Learn More
Raybiotech Inc Mouse IL-36 beta/IL-1F8 ELISA Kit
Supplier:  Raybiotech Inc ELMIL36B1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 6.5 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Mouse IL-36 beta.
Catalog No. 50-195-0055