Learn More
Sino Biological Mouse IL-4 ELISpot Kit (ALP) (Strips) 1 Kit (96 Tests)
Catalog No. 5000696246
Encompass_Preferred
The kit applies for the detection of mouse IL-4 at the cellular level by counting IL-4-secreting cells.
Catalog No. 50-006-96246 Supplier Sino Biological Supplier No. EK51084B1
The kit applies for the detection of mouse IL-4 at the cellular level by counting IL-4-secreting cells.