Learn More
Mouse IL-4 ELISPOT Pair, BD
Catalog No. BDB551878 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
5 x 96-well Plates
Catalog No. Quantity
BDB551878 5 x 96-well Plates
1 options
Catalog No. BDB551878 Supplier BD Biosciences Supplier No. 551878
Add to Cart
Add to Cart
Includes
Mouse IL-4 ELISPOT Capture Antibody (no azide/low endotoxin), Mouse IL-4 ELISPOT Detection Antibody (biotinylated) and a Certificate of Analysis that provides lot-specific optimal reagent concentrations
Convenient set of unlabelled anti-IL-4 capture antibody and a biotinylated anti-IL-4 detection antibody
TRUSTED_SUSTAINABILITY
Specifications
Quantity 5 x 96-well Plates