Learn More
FUJIFILM BIOSCIENCES INC Mouse IL-4, Recombinant Protein, Shenandoah Biotechnology, 20 ug vial
Supplier:  FUJIFILM BIOSCIENCES INC 2001820UG
Encompass_Preferred
Interleukin 4 (IL-4) is an immunomodulatory cytokine that functions to induce naïve helper T cells to differentiate into type 2 T helper (Th2) cells. Th2 cells subsequently produce more IL-4 in a positive feedback loop. IL-4 also promotes immunoglobulin IgG to IgE isotype switching on B cells. IL-4 binds the IL-4RA receptor to activate STAT6 signaling.
Catalog No. 50-990-850