Learn More
Invitrogen™ Mouse IL-5 ProQuantum Immunoassay Kit

Description
Specifications
Specifications
| Accession Number | P04401 |
| Assay Range | 0.32-5000 pg/mL |
| Assay Sensitivity | <0.3 pg/mL |
| Conjugate | FAM |
| Detection Method | Taqman™ Real-time PCR |
| Immunoassay Kit Format | Proximity-based Immunoassay |
| Product Type | ProQuantum |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Mouse |
| For Use With (Application) | Protein Quantitation |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.