Learn More
Sino Biological Mouse IL-6 Matched ELISA Antibody Pair Set 15 Plates
Catalog No. 5000786318
Encompass_Preferred
The Mouse IL-6 ELISA Pair Set is for the quantitative determination of Mouse IL-6.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Mouse IL-6 coated on a 96-well plate. Standards and samples are added to the wells, and any Mouse IL-6 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated Rabbit anti-Mouse IL-6 monoclonal antibody is then added, producing an antibody-antigen-antibody ""sandwich"". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Mouse IL-6 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86318 Supplier Sino Biological Supplier No. SEK5013615
The Mouse IL-6 ELISA Pair Set is for the quantitative determination of Mouse IL-6.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Mouse IL-6 coated on a 96-well plate. Standards and samples are added to the wells, and any Mouse IL-6 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated Rabbit anti-Mouse IL-6 monoclonal antibody is then added, producing an antibody-antigen-antibody ""sandwich"". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Mouse IL-6 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.