Learn More
ACROBiosystems Mouse IL-6 Protein, His Tag
Supplier:  ACROBiosystems IL6M524550UG
Encompass_Preferred
Recombinant Protein;50UG;Mouse IL-6, His Tag (IL6-M5245) is expressed from human 293 cells (HEK293). It contains AA Phe 25 - Thr 211 (Accession # P08505-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 23.6 kDa. The protein migrates as 25-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse Human IL-6, His Tag (Cat. No. IL6-M5245) at 5 μg/mL (100 μL/well) can bind Biotinylated Mouse IL-6 R alpha, His,Avitag (Cat. No. ILR-M82E9) with a linear range of 20-156 ng/mL (QC tested).
Catalog No. 50-205-4823