Learn More
Description
Mature mouse IL-6 was expressed in Pichia pastoris, X-33. GenBank Accession # NP_112445, amino acids 25 - 211. For protein verification, the expressed protein was N-terminal sequenced and begins with the following 10 amino acids: FPTSQVRRGD. Purity is >95% by SDS-PAGE analyses. Use: For in vitro applications. A dose-response assay should be performed to determine the optimal concentration for specific experiments. Reconstitution and Dilution: Store lyophilized material at 4°C. Reconstitute in 0.5 mL of distilled or deionized water. Dilute in a buffer-containing carrier protein, aliquot, and store at or below -70°C, avoiding multiple freeze-thaw cycles. PBS with 1% BSA or a neutral media containing <10% FCS are suitable buffers.
Specifications
Specifications
| Accession Number | P08505 |
| For Use With (Application) | Bioactivity, ELISA, Immunoprecipitation, Western Blot |
| Formulation | PBS with no preservative |
| Gene ID (Entrez) | 16193 |
| Molecular Weight (g/mol) | 21.7 kDa |
| Name | Mouse IL-6 |
| Purification Method | Purified |
| Quantity | 20 μg |
| Source | Yeast |
| Storage Requirements | 4°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.