Learn More
Bon Opus Biosciences Recombinant Mouse IL-6R (C-mFc) 50ug
SDP
Supplier:  Bon Opus Biosciences BP107950UG
Encompass_Preferred
Recombinant Mouse IL-6R (C-mFc) is produced by Human Cells. The target gene encoding Leu20-Glu357 is expressed with a C-mFc tag. Accession# P22272-1. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1079
Catalog No. 50-217-2677