Learn More
FUJIFILM BIOSCIENCES INC Mouse IL-7, Recombinant Protein, Shenandoah Biotechnology, 10 ug vial
Supplier:  FUJIFILM BIOSCIENCES INC 2000310UG
Encompass_Preferred
Interleukin 7 (IL-7) is a hematopoietic cytokine that is an important regulator of B and T cell development. IL-7 is secreted by bone marrow and thymic stromal cells, dendritic cells, intestinal epithelial cells, hepatocytes, and keratinocytes. IL-7 signals through the interleukin 7 receptor (IL-7R) to promote the differentiation of hematopoietic stem cells into T cells, B cells, and natural killer cells. IL-7 is also a regulator of intestinal mucosal lymphocyte proliferation. Human and mouse IL-7 show species cross-reactivity.
Catalog No. 50-592-294
Edge