Learn More
ACROBiosystems Mouse IL-7 R alpha / CD127 Protein, Fc Tag
Supplier:  ACROBiosystems ILRM5253100UG
Encompass_Preferred
Mouse IL-7 R alpha Protein, Fc Tag (ILR-M5253) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Asp 239 (Accession # Q9R0C1).
Catalog No. 50-233-0898