Learn More
Proteintech Group Inc Mouse IL1-alpha ELISA 96well
SDP
Supplier:  Proteintech Group Inc KE1002496T
Encompass_Preferred
Interleukin-1α (Il-1α) is a protein that plays a crucial role in the regulation of a number of key cellular processes. Il-1α is a cytokine and a potent mediator of body’s response to inflammation, microbial invasion, tissue injury and immunological response. In addition, recent studies suggest that Il-1α also has a role in wound healing, rheumatoid arthritis, Alzheimer’s disease and tumor growth. Il-1α functions as an alarmin during the process of sterile inflammation. It serves as a signal from dying cells to potentiate the inflammatory response, increasing the expression of other secreted factors like IL-6 and IL-8. Il-1α was found to affect the cell cycle of osteosarcoma cell to reduce cell growth.
Catalog No. 50-197-4148