Learn More
Proteintech Group Inc Mouse IL1-beta ELISA 96well
SDP
Supplier:  Proteintech Group Inc KE1000396T
Encompass_Preferred
Interleukin-1 is a pro-inflammatory cytokine with multiple biological effects. The IL-1 gene family encodes three proteins: IL-1α, IL-1β and their naturally occurring inhibitor IL-1RN. Interleukin 1β(IL-1β), mainly produced by blood monocytes and tissue macrophages, has been implicated in mediating both acute and chronic inflammation. IL-1β is known to be involved in a variety of cellular activities, including cell proliferation, differentiation and apoptosis. IL-1β is emerging as a key mediator of carcinogenesis that characterizes host-environment interactions.
Catalog No. 50-197-4129