Learn More
Sino Biological Recombinant Anti-IL-10RB/IL-10R beta Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50846R00920
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL10RB / IL-10RB (rM IL10RB / IL-10RB; Catalog#50846-M08H; NP_032375.2; Met1-Ser222).
Catalog No. 50-255-3059