Learn More
Sino Biological Anti-IL-12 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological CT022RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse IL-12 (IL12A & IL12B Heterodimer) (rM IL-12 (IL12A & IL12B Heterodimer); Catalog#CT022-M08H; P43431 & P43432; Met1-Ala215 & Met1-Ser335). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-7816