Learn More
Sino Biological Recombinant Mouse IL-12 Protein (His & hFc Tag)
Supplier:  Sino Biological CT024M0208H1
Encompass_Preferred
A DNA sequence encoding the p35 subunit of mouse IL12, termed as IL12A (P43431) (Met 1-Ala 215) was fused with the Fc region of human IgG1 at the C-terminus, constructed the plasmid 1; A DNA sequence encoding the p40 subunit of mouse IL12, termed as IL12B (P43432) (Met 1-Ser 335) was fused with a polyhistidine tag at the C-terminus, constructed the plasmid 2. The two plasmids were co-expressed and the heterodimer was purified.
Catalog No. 50-255-2622