Learn More
Sino Biological Anti-IL-23 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological CT028T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse IL-23 (IL23A & IL12B Heterodimer) (rM IL-23 (IL23A & IL12B Heterodimer); Catalog#CT028-M08H; Q9EQ14 & P43432; Met1-Ala196 & Met1-Ser335). IL-23 (IL23A & IL12B Heterodimer) specific IgG was purified by Mouse IL-23 (IL23A & IL12B Heterodimer) affinity chromatography.
Catalog No. 50-240-9066