Learn More
Sino Biological Recombinant Anti-IL-12 p40/IL12B Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 51004R020100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL12B / IL-12B (Catalog#51004-M08H; NP_001290173.1; Met1-Ser335).
Catalog No. 50-241-1348