Learn More
Sino Biological Recombinant Mouse IL-23 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological CT140M08H50
Encompass_Preferred
A DNA sequence encoding the Mouse IL12B (P43432)(Met1-Ser335) was expressed, constructed the plasmid 1; A DNA sequence encoding the Mouse IL23A (Q9EQ14) (Met1-Ala196) was expressed with a polyhistidine tag at the C-terminus, constructed the plasmid 2. The two plasmids were co-expressed and the IL12B & IL23A heterodimer was purified.
Catalog No. 50-255-2588