Learn More
Sino Biological Recombinant Mouse IL-12 R beta 2/IL12RB2 Protein (His Tag), HPLC-verified 1
Supplier:  Sino Biological 50099M08H1
Encompass_Preferred
A DNA sequence encoding the mouse IL12RB2 (NP_032380.1) extracellular domain (Met 1-Asn 637) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6670