Learn More
Sino Biological Recombinant Anti-IL-12 R beta 2/IL12RB2 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50099R093100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL12RB2 (rM IL12RB2; Catalog#50099-M08H; NP_032380.1; Met1-Asn637).
Catalog No. 50-255-3465