Learn More
Sino Biological Anti-IL-12 R beta 2/IL12RB2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50099T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse IL12RB2 (Catalog#50099-M08H; NP_032380.1; Met1-Asn637). IL12RB2 specific IgG was purified by Mouse IL12RB2 affinity chromatography.
Catalog No. 50-255-3463