Learn More
Sino Biological Recombinant Mouse IL-13 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50225M08H500
Encompass_Preferred
A DNA sequence encoding the Mouse IL13 (P20109) (Met1-Phe131) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-299-3365