Learn More
Sino Biological Recombinant Mouse IL-13 Protein (His Tag), HPLC-verified
Catalog No. 502993311
Encompass_Preferred
A DNA sequence encoding the Mouse IL13 (P20109) (Met1-Phe131) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-299-3311 Supplier Sino Biological Supplier No. 50225M08H50
A DNA sequence encoding the Mouse IL13 (P20109) (Met1-Phe131) was expressed with a polyhistidine tag at the C-terminus.